missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15480759
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
694501-34-3
C12H16FN3O2
253.277
XMJXGMZBNFICRS-UHFFFAOYSA-N
2947839
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F
This item is not returnable. View return policy
694501-34-3
C12H16FN3O2
253.277
XMJXGMZBNFICRS-UHFFFAOYSA-N
2947839
CCN1CCN(CC1)C2=C(C=C(C=C2)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Beilstein 15470780
Synonym 1-ethyl-4-2-fluoro-4-nitrophenyl piperazine, 1-ethyl-4-2-fluoro-4-nitro-phenyl-piperazine, piperazine, 1-ethyl-4-2-fluoro-4-nitrophenyl, piperazine,1-ethyl-4-2-fluoro-4-nitrophenyl
Formula Weight 253.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Ethyl-4-(2-fluoro-4-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'