missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium bromide, 98%, Thermo Scientific Chemicals

Catalog Number p-3736476
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
65039-08-9
C6H11BrN2
191.07
MFCD03427610
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
2734235
[Br-].CCN1C=C[N+](C)=C1
This item is not returnable. View return policy
65039-08-9
C6H11BrN2
191.07
MFCD03427610
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium bromide, 3-ethyl-1-methyl-1h-imidazol-3-ium bromide, 1-ethyl-3-methylmidazolium bromide, unii-xo254ye73i, 1-ethyl-3-methyl-1h-imidazol-3-ium bromide, acmc-1b3a0, dsstox_cid_29300, dsstox_rid_83418, dsstox_gsid_49343, c6h11n2.br
2734235
[Br-].CCN1C=C[N+](C)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
IUPAC Name 1-ethyl-3-methyl-1H-imidazol-3-ium bromide
Formula Weight 191.07
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1-Ethyl-3-methylimidazolium bromide
Show More Show Less
compliance-icons
Product Identifier
  • 1-Ethyl-3-methylimidazolium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'