missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-3-hydroxypyridinium bromide, 98%

Brand:  Alfa Aesar™ H55169.22

Code : NEW

Additional Details : CAS Number : 80866-84-8 Weight : 0.10000kg Transport : UN number : 3263 Chem class : 1b Pack group : II

Quantity 100g

 View more versions of this product

Product Code. 15481328

  • £444.56 / 100g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

1-Ethyl-3-hydroxypyridinium bromide
204.07
UN3263
80866-84-8
MFCD00011995

Specifications

1-Ethyl-3-hydroxypyridinium bromide
104° to 108°C
80866-84-8
98%
MFCD00011995
White to pale brown
100g
Solid
204.07
UN3263
Videos
SDS