missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
Specifications
Color | Yellow |
Quantity | 50 g |
Sensitivity | Moisture sensitive |
Formula Weight | 212 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 1-Ethyl-2,3-dimethylimidazolium tetrafluoroborate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'