missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Ethyl-1-methylpyrrolidinium bromide, 98%

Brand:  Alfa Aesar™ H66050.22

Code : NEW

Additional Details : CAS Number : 69227-51-6 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15462689

  • £113.70 / 100g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Description

Description

Applications
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications

Chemical Identifiers

1-Ethyl-1-methylpyrrolidinium bromide
194.12
14507266
69227-51-6
MFCD03095384

Specifications

1-Ethyl-1-methylpyrrolidinium bromide
69227-51-6
98%
MFCD03095384
100g
Solid
194.12
14507266
Videos
SDS