missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethoxy-2-propanol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -100°C |
Color | Colorless |
Density | 0.8970g/mL |
Boiling Point | 132°C |
Flash Point | 42°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. Remark: balance 2-ethoxy-1-propanol (GC) |
Packaging | Glass Bottle |
Linear Formula | C2H5OCH2CH(OH)CH3 |
Refractive Index | 1.4050 to 1.409 |
Show More |
Safety and Handling
Product Identifier
- 1-Ethoxy-2-propanol
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'