missing translation for 'onlineSavingsMsg'
Learn More

1-EBIO, Tocris Bioscience™

Activator of epithelial KCa channels

Brand:  Tocris (Bio-Techne) 1041/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13448363

  • £324.87 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Activator of epithelial KCa channels, stimulates a large and sustained trans-epithelial Cl- secretory response across T84 monolayers. Induces hyperpolarization to the same magnitude as ACh in aortic value endothelial cells. Promotes embryonic stem cell (ESC) differentiation into cardiomyocytes.
Specifications

Chemical Identifiers

10045-45-1
162.192
1-ethyl-2-benzimidazolinone, 1-ebio, 1-ethylbenzimidazolinone, 1-ethyl-1h-benzo d imidazol-2 3h-one, 1-ethylbenzimidazolin-2-one, 1-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-ethyl-benzimidazolinone, unii-m82w79ss4w, 2h-benzimidazol-2-one, 1-ethyl-1,3-dihydro, ebio
CHEBI:34076
CCN1C2=CC=CC=C2NC1=O
C9H10N2O
CXUCKELNYMZTRT-UHFFFAOYSA-N
82320
3-ethyl-1H-benzimidazol-2-one

Specifications

50mg
10045-45-1
1-ethyl-2-benzimidazolinone, 1-ebio, 1-ethylbenzimidazolinone, 1-ethyl-1h-benzo d imidazol-2 3h-one, 1-ethylbenzimidazolin-2-one, 1-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-ethyl-benzimidazolinone, unii-m82w79ss4w, 2h-benzimidazol-2-one, 1-ethyl-1,3-dihydro, ebio
CCN1C2=CC=CC=C2NC1=O
162.192
CHEBI:34076
>99%
1-EBIO
C9H10N2O
CXUCKELNYMZTRT-UHFFFAOYSA-N
3-ethyl-1H-benzimidazol-2-one
82320
162.19
Videos
Documents