missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyl isocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-7046180
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4202-38-4
C13H25NO
211.349
MFCD00041744
YIDSTEJLDQMWBR-UHFFFAOYSA-N
dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, isocyanic acid dodecyl ester
77873
1-isocyanatododecane
CCCCCCCCCCCCN=C=O
This item is not returnable. View return policy
4202-38-4
C13H25NO
211.349
MFCD00041744
YIDSTEJLDQMWBR-UHFFFAOYSA-N
dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, isocyanic acid dodecyl ester
77873
1-isocyanatododecane
CCCCCCCCCCCCN=C=O

1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Density 0.877
Boiling Point 157°C to 161°C (21 mmHg)
Flash Point 134°C (273°F)
Odor Irritating
Refractive Index 1.441
Quantity 1 g
UN Number UN2206
Beilstein 1774823
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 211.35
Percent Purity 98%
Chemical Name or Material 1-Dodecyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'