missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxynojirimycin, Tocris Bioscience™

Glucosidase I and II inhibitor

Brand:  Tocris (Bio-Techne) 12585

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13429003

  • £119.07 / 5mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of glucosidase I (Ki = 2.1 mM) and II (Ki = 7 mM).
Specifications

Chemical Identifiers

19130-96-2
163.173
(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
CHEBI:44369
C1C(C(C(C(N1)CO)O)O)O
C6H13NO4
LXBIFEVIBLOUGU-JGWLITMVSA-N
29435
(2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol

Specifications

1-Deoxynojirimycin
C6H13NO4
LXBIFEVIBLOUGU-JGWLITMVSA-N
(2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
29435
163.17
5mg
19130-96-2
(2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
C1C(C(C(C(N1)CO)O)O)O
163.173
CHEBI:44369
>98%
Videos
Documents