Learn More
Tocris Bioscience™ 1-Deoxymannojirimycin hydrochloride
Brand:  Tocris Bioscience™ 125925
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13459003
  • £368.29 / 25mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Selective inhibitor of α-mannosidase I.
Specifications
Specifications
(2R,3R,4R,5R)-2-(Hydroxymethyl)-3,4,5-piperidinetriol hydrochloride
C6H13NO4.HCl
25mg
73465-43-7
Observed MW: 199.63
Soluble to 50mM in water