missing translation for 'onlineSavingsMsg'
Learn More

1-Decyne, 98%, Thermo Scientific Chemicals

Catalog Number 15159253
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
764-93-2
C10H18
138.25
MFCD00009576
ILLHQJIJCRNRCJ-UHFFFAOYSA-N
1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
12997
CHEBI:87322
dec-1-yne
CCCCCCCCC#C
764-93-2
C10H18
138.25
MFCD00009576
ILLHQJIJCRNRCJ-UHFFFAOYSA-N
1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
12997
CHEBI:87322
dec-1-yne
CCCCCCCCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -44.0°C
Color Colorless to Yellow
Density 0.7600g/mL
Boiling Point 174.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7C≡CH
Refractive Index 1.4260 to 1.4280
Quantity 100 mL
Beilstein 01,I,122
Specific Gravity 0.76
Formula Weight 138.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Decyne, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Decyne
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'