missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Decylphosphonic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 100°C to 101°C |
Quantity | 1 g |
UN Number | UN3261 |
Beilstein | 1771572 |
Formula Weight | 222.27 |
Percent Purity | 98% |
Chemical Name or Material | 1-Decylphosphonic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'