missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1-Decyl)triphenylphosphonium bromide, 98%

Brand:  Alfa Aesar™ A11021.14

Code : NEW

Additional Details : CAS Number : 32339-43-8 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11464300

  • £17.80 / 25g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

(1-Decyl)triphenylphosphonium bromide
CH3(CH2)9P(C6H5)3Br
MFCD00051854
32339-43-8
483.48
4076987

Specifications

(1-Decyl)triphenylphosphonium bromide
98%
483.48
≈90°C
25g
32339-43-8
CH3(CH2)9P(C6H5)3Br
MFCD00051854
4076987
Videos
SDS