missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (1-Decyl)triphenylphosphonium bromide, 98%

Brand:  Alfa Aesar™ A11021.22

Code : NEW

Additional Details : CAS Number : 32339-43-8 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11474300

  • £69.50 / 100g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

(1-Decyl)triphenylphosphonium bromide
CH3(CH2)9P(C6H5)3Br
MFCD00051854
32339-43-8
483.48
4076987

Specifications

(1-Decyl)triphenylphosphonium bromide
98%
483.48
≈90°C
100g
32339-43-8
CH3(CH2)9P(C6H5)3Br
MFCD00051854
4076987
Videos
SDS