missing translation for 'onlineSavingsMsg'
Learn More

1-Decene, ca. 95%, Thermo Scientific Chemicals

Catalog Number p-4258922
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
872-05-9
C10H20
140.27
AFFLGGQVNFXPEV-UHFFFAOYSA-N
1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
13381
CHEBI:87315
dec-1-ene
CCCCCCCCC=C
This item is not returnable. View return policy
872-05-9
C10H20
140.27
AFFLGGQVNFXPEV-UHFFFAOYSA-N
1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
13381
CHEBI:87315
dec-1-ene
CCCCCCCCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point -66.0°C
Color Colorless
Density 0.7410g/mL
Boiling Point 169.0°C to 171.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)7CH=CH2
Refractive Index 1.4200 to 1.4220
Quantity 1 L
Beilstein 01,III,858
Specific Gravity 0.741
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Viscosity 1.1 mm2/s (20°C)
Formula Weight 140.27
Percent Purity ca. 95%
Physical Form Liquid
Chemical Name or Material 1-Decene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Decene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'