missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Decanol, 98+%, Thermo Scientific Chemicals
17.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mL
2500 mL
Description
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
1-Decanol is used in the production of plasticizers, lubricants, surfactants and solvents. It is also used to study the thermal properties of polymer-monolithic stationary phases. Further, it is used to enhance homomeric glycine receptor function. In addition to this, it is used in daily flavor, food flavor and cosmetics.
Solubility
Soluble in alcohol, ether. Insoluble in water.Miscible with carbon tetrachloride, ethanol, ether, acetone, benzene, chloroform and glacial acetic acid. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications
Melting Point | 6°C |
Density | 0.828 g/mL |
Boiling Point | 230°C to 232°C |
Flash Point | 82°C (179°F) |
Assay Percent Range | >98% |
Linear Formula | CH3(CH2)8CH2OH |
Refractive Index | 1.437 |
Quantity | 500 mL |
UN Number | UN3082 |
Beilstein | 1735221 |
Show More |
Safety and Handling
Product Identifier
- 1-Decanol
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1-Decanol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'