missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11926231
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1233526-60-7
C14H23BN2O2
262.16
OCBNABJVDPSMQY-UHFFFAOYSA-N
1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
56776730
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3
This item is not returnable. View return policy
1233526-60-7
C14H23BN2O2
262.16
OCBNABJVDPSMQY-UHFFFAOYSA-N
1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
56776730
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3
Melting Point 38°C to 40°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 262.16
Percent Purity 97%
Chemical Name or Material 1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'