missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentene-1-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3770934
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1560-11-8
C6H8O2
112.13
MFCD00001396
PYRZPBDTPRQYKG-UHFFFAOYSA-N
1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
95964
cyclopentene-1-carboxylic acid
C1CC=C(C1)C(=O)O
This item is not returnable. View return policy
1560-11-8
C6H8O2
112.13
MFCD00001396
PYRZPBDTPRQYKG-UHFFFAOYSA-N
1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
95964
cyclopentene-1-carboxylic acid
C1CC=C(C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 121.0°C to 124.0°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C5H7COOH
Quantity 1 g
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 1-Cyclopentene-1-carboxylic acid
compliance-icons
Product Identifier
  • 1-Cyclopentene-1-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'