missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclohexylmethyl)piperazine, 97%, Thermo Scientific™

Catalog Number 15431768
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
57184-23-3
C11H22N2
182.31
MFCD01075237
LRPGNFROBDUREU-UHFFFAOYSA-N
2735885
1-(cyclohexylmethyl)piperazine
C(C1CCCCC1)N1CCNCC1
This item is not returnable. View return policy
57184-23-3
C11H22N2
182.31
MFCD01075237
LRPGNFROBDUREU-UHFFFAOYSA-N
2735885
1-(cyclohexylmethyl)piperazine
C(C1CCCCC1)N1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.938 g/mL
Boiling Point 230°C to 231°C
Flash Point 87°C (189°F)
Refractive Index 1.495
Quantity 250 mg
Synonym 1-cyclohexylmethyl piperazine, piperazine, 1-cyclohexylmethyl, cyclohexylmethyl piperazine, 1-cyclohexylmethyl-piperazine, 1-cyclohexyl-methyl piperazine, piperazine,1-cyclohexylmethyl
Formula Weight 182.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(Cyclohexylmethyl)piperazine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'