missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexenylacetonitrile, 90%, Thermo Scientific™

Catalog Number 10666292
Click to view available options
10g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 6975-71-9
CAS Min % 88.0
CAS Max % 100.0
Density 0.9400g/mL
Boiling Point 144°C (90.0mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C8H11N
Linear Formula C6H9CH2CN
Refractive Index 1.4780 to 1.48
MDL Number MFCD00001566
Quantity 10g
Beilstein 09,46
Specific Gravity 0.94
Synonym 1-cyclohexene-1-acetonitrile, 2-cyclohex-1-en-1-yl acetonitrile, 1-cyclohexenylacetonitrile, cyclohexenylacetonitrile, cyclohex-1-enyl-acetonitrile, 1-cyclohexen-1-ylacetonitrile, 1-cyclohexenacetonitrile, 1-cyclohexeneacetonitrile, 2-cyclohexen-1-yl acetonitrile, 2-cyclohex-1-enylethanenitrile
InChI Key OYEXEQFKIPJKJK-UHFFFAOYSA-N
SMILES N#CCC1=CCCCC1
IUPAC Name 2-(cyclohexen-1-yl)acetonitrile
Molecular Weight (g/mol) 121.18
PubChem CID 81461
Formula Weight 121.18
Percent Purity 90%
Chemical Name or Material 1-Cyclohexenylacetonitrile, 92%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Cyclohexenylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'