missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Cyclohexenyl trifluoromethanesulfonate, 97%

Brand:  Alfa Aesar™ H55879.03

Code : NEW

Additional Details : CAS Number : 28075-50-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462268

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

Applications
Used as a pharmaceutical intermediate.

Notes
Store at 4°C.
Specifications

Chemical Identifiers

1-Cyclohexenyl trifluoromethanesulfonate
28075-50-5
230.2
MFCD06411281

Specifications

1-Cyclohexenyl trifluoromethanesulfonate
Liquid
230.2
1.315g/mL
Colorless to yellow
28075-50-5
97%
MFCD06411281
84° to 87°C (4mmHg)
1g
Videos
SDS