Alfa Aesar™ 1-Cyclohexenyl trifluoromethanesulfonate, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H55879.14

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 28075-50-5 Weight: 0.02500kg


Quantity 25g

Product Code. 15482268

Quantity Price
1 £ 468.52 / 25g
Estimated Shipment date
from Supplier 12-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-Cyclohexenyl trifluoromethanesulfonate
CAS 28075-50-5
Formula Weight 230.2
MDL Number MFCD06411281

Specification

Formula Weight 230.2
Boiling Point 84° to 87°C (4mmHg)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
CAS 28075-50-5
Assay 97%
Quantity 25g
MDL Number MFCD06411281
Health Hazard 1 Warning
Chemical Name or Material 1-Cyclohexenyl trifluoromethanesulfonate
Density 1.315g/mL
Color Colorless to yellow
Physical Form Liquid

Applications
Used as a pharmaceutical intermediate.

Notes
Store at 4°C.

Documentation

Technical documents

Safety and Handling

Warning

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.