missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanomethylpiperidine, 98%, Thermo Scientific™

Catalog Number p-7020486
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
3010-03-5
MFCD00023733
This item is not returnable. View return policy
3010-03-5
MFCD00023733

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼25°C
Density 0.96
Boiling Point 210°C
Molecular Formula C12H9BrClNO3
Refractive Index 1.466
Quantity 1 g
UN Number UN3439
Synonym 1-acetyl-5-bromo-4-chloro-1h-indol-3-yl acetate, 5-bromo-4-chloroindoxyl 1,3-diacetate, 5-bromo-4-chloroindoxyl-1,3-diacetate, 3-acetoxy-1-acetyl-5-bromo-4-chloroindole, 1-acetyl-5-bromo-4-chloro-1h-indol-3-ylacetate, 1h-indol-3-ol, 1-acetyl-5-bromo-4-chloro-, acetate ester, acmc-1ag9t, 5-bromo-4-chloroindolyl 1,3-diacetate, 5-bromo-4-chloro-indoxyl-1,3-diacetate, cas
InChI Key DSHQTSIXXYZXGR-UHFFFAOYSA-N
SMILES CC(=O)N1C=C(C2=C1C=CC(=C2Cl)Br)OC(=O)C
IUPAC Name (1-acetyl-5-bromo-4-chloroindol-3-yl) acetate
Molecular Weight (g/mol) 330.56
PubChem CID 76416
Formula Weight 124.19
Percent Purity 98%
Chemical Name or Material 1-Cyanomethylpiperidine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'