missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Description
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.
Solubility
Soluble in water, and acetonitrile.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate is a cyanylation reagent for protein sulfhydryl groups and used to prepare protein-polysaccharide conjugates. It is employed as an activating agent for polysaccharide resins and conjugation of lipopolysaccharides while retaining endotoxic activity.
Solubility
Soluble in water, and acetonitrile.
Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Specifications
Melting Point | 196°C to 200°C |
Quantity | 100 mg |
Beilstein | 5685616 |
Solubility Information | Soluble in water,and acetonitrile. |
Formula Weight | 234.99 |
Percent Purity | 98% |
Chemical Name or Material | 1-Cyano-4-(dimethylamino)pyridinium tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'