missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(chloromethyl)-4-(trifluoromethoxy)benzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH2ClC6H4OCF3 |
Quantity | 1 g |
Formula Weight | 210.58 |
Percent Purity | 97% |
Chemical Name or Material | 1-(Chloromethyl)-4-(trifluoromethoxy)benzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'