missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroisoquinoline, 97+%, Thermo Scientific Chemicals

Catalog Number p-3593222
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19493-44-8
C9H6ClN
163.61
MFCD00024134
MSQCQINLJMEVNJ-UHFFFAOYSA-N
1-chloro-isoquinoline, isoquinoline, 1-chloro, chloroisoquinoline, isoquinoline, chloro, 1-chloroisoquinone, 1-chloro isoquinoline, zlchem 1129, pubchem13906, acmc-1bowu, 1-chloroisoquinoline
140539
1-chloroisoquinoline
C1=CC=C2C(=C1)C=CN=C2Cl
This item is not returnable. View return policy
19493-44-8
C9H6ClN
163.61
MFCD00024134
MSQCQINLJMEVNJ-UHFFFAOYSA-N
1-chloro-isoquinoline, isoquinoline, 1-chloro, chloroisoquinoline, isoquinoline, chloro, 1-chloroisoquinone, 1-chloro isoquinoline, zlchem 1129, pubchem13906, acmc-1bowu, 1-chloroisoquinoline
140539
1-chloroisoquinoline
C1=CC=C2C(=C1)C=CN=C2Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Melting Point 31°C to 36°C
Color White to Yellow
Boiling Point 274°C to 275°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.61
Percent Purity 97+%
Physical Form Low Melting Solid, Crystals and/or Chunks
Chemical Name or Material 1-Chloroisoquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'