missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloroethyl chloroformate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
Specifications
CAS Min % | 97.0 |
CAS Max % | 100.0 |
Color | Colorless |
Density | 1.3200g/mL |
Boiling Point | 118°C to 119°C |
Flash Point | 40°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% min. (GC) |
Packaging | Glass bottle |
Linear Formula | ClCO2CH(Cl)CH3 |
Show More |
Safety and Handling
Product Identifier
- 1-Chloroethyl chloroformate
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H226-Flammable liquid and vapour.
- EUH029-Contact with water liberates toxic gas.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P402+P404-Store in a dry place. Store in a closed container.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'