missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane, 99+%, pure, Thermo Scientific Chemicals
146.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Melting Point | -123.0°C |
Color | Colorless |
Density | 0.8800g/mL |
Boiling Point | 77.0°C to 78.0°C |
Flash Point | −12°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Plastic drum |
Show More |
Safety and Handling
Product Identifier
- 1-Chlorobutane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- H304-May be fatal if swallowed and enters airways.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P331-Do NOT induce vomiting.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'