missing translation for 'onlineSavingsMsg'
Learn More

1-Chloroadamantane, 98%, Thermo Scientific Chemicals

Catalog Number p-7049581
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
935-56-8
C10H15Cl
170.68
MFCD00075627
OZNXTQSXSHODFR-UHFFFAOYSA-N
1-adamantyl chloride, adamantyl chloride, 3-chloroadamantane, adamantane, 1-chloro, tricyclo 3.3.1.13,7 decane, 1-chloro, 1-chlorotricyclo 3.3.1.13,7 decane, 3r,5s,7s-1-chloroadamantane, 1-chlorotricyclo 3.3.1.1∼3,7∼ decane
64154
1-chloroadamantane
ClC12CC3CC(CC(C3)C1)C2
This item is not returnable. View return policy
935-56-8
C10H15Cl
170.68
MFCD00075627
OZNXTQSXSHODFR-UHFFFAOYSA-N
1-adamantyl chloride, adamantyl chloride, 3-chloroadamantane, adamantane, 1-chloro, tricyclo 3.3.1.13,7 decane, 1-chloro, 1-chlorotricyclo 3.3.1.13,7 decane, 3r,5s,7s-1-chloroadamantane, 1-chlorotricyclo 3.3.1.1∼3,7∼ decane
64154
1-chloroadamantane
ClC12CC3CC(CC(C3)C1)C2

It is used in some dry etching masks and polymer formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 165°C
Quantity 5 g
Solubility Information Poorly soluble in water. Soluble in hydrocarbons. But readily soluble in nonpolar organic solvents.
Formula Weight 170.68
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 1-Chloroadamantane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'