missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-8-hydroxyquinoline, 95%, Thermo Scientific Chemicals

Catalog Number 10590171
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
130-16-5
C9H6ClNO
179.61
MFCD00006788
CTQMJYWDVABFRZ-UHFFFAOYSA-N
5-chloro-8-hydroxyquinoline, cloxiquine, cloxyquin, 5-chloro-8-quinolinol, dermofongin, 5-chlorooxine, 8-quinolinol, 5-chloro, chlorisept, dermofungin, 5-chloro-8-oxyquinoline
2817
5-chloroquinolin-8-ol
C1=CC2=C(C=CC(=C2N=C1)O)Cl
This item is not returnable. View return policy
130-16-5
C9H6ClNO
179.61
MFCD00006788
CTQMJYWDVABFRZ-UHFFFAOYSA-N
5-chloro-8-hydroxyquinoline, cloxiquine, cloxyquin, 5-chloro-8-quinolinol, dermofongin, 5-chlorooxine, 8-quinolinol, 5-chloro, chlorisept, dermofungin, 5-chloro-8-oxyquinoline
2817
5-chloroquinolin-8-ol
C1=CC2=C(C=CC(=C2N=C1)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 121°C to 126°C
Color Gray to Green
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Quantity 500 g
Beilstein 21, 95
Merck Index 15, 2405
Formula Weight 179.61
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Chloro-8-hydroxyquinoline, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Chloro-8-hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'