missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
Specifications
Refractive Index | 1.525 |
Quantity | 10 g |
Beilstein | 1732408 |
Sensitivity | Light sensitive |
Solubility Information | Not miscible in water. |
Formula Weight | 246.52 |
Percent Purity | 97% |
Chemical Name or Material | 1-Chloro-6-iodohexane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'