missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-6-hydroxyhexane, 95%, ACROS Organics™

Brand:  Acros Organics 109285000

Code : 29

Additional Details : CAS Number : 2009-83-8 Weight : 0.55000kg Transport : UN number : 2810 Chem class : 1 Pack group : III

Quantity 500g
Packaging Glass bottle

 View more versions of this product

Product Code. 10761424

  • £192.40 / 500g


Estimated Shipment
18-05-2018
Add to basket

Specifications

Chemical Identifiers

1-chloro-6-hydroxyhexane
94.0
95%
C6H13ClO
136.62
6-chloro-1-hexanol, Hexamethylene chlorohydrin
2009-83-8
100.0
95%
Cl(CH2)6OH
MFCD00002984

Specifications

1-chloro-6-hydroxyhexane
98°C
Clear colorless to pale yellow
Authentic
94.0
Glass bottle
Liquid
95%
Cl(CH2)6OH
MFCD00002984
1.0300g/mL
108.0° to 112.0°C (14.0 mmHg)
500g
2009-83-8
100.0
1.4547 to 1.4567
95%
C6H13ClO
136.62
6-chloro-1-hexanol, Hexamethylene chlorohydrin
Videos
SDS