missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-iodobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-88429
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
637-87-6
C6H4ClI
238.46
MFCD00001053
GWQSENYKCGJTRI-UHFFFAOYSA-N
4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
12510
1-chloro-4-iodobenzene
C1=CC(=CC=C1Cl)I
This item is not returnable. View return policy
637-87-6
C6H4ClI
238.46
MFCD00001053
GWQSENYKCGJTRI-UHFFFAOYSA-N
4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
12510
1-chloro-4-iodobenzene
C1=CC(=CC=C1Cl)I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 52°C to 55°C
Color White to Beige
Boiling Point 226°C to 227°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4I
Quantity 25 g
Beilstein 05, 221
Formula Weight 238.46
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Chloro-4-iodobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-4-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'