missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-iodobenzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Specifications
Density | 1.9200g/mL |
Assay Percent Range | 97% |
Linear Formula | ClC6H4I |
Quantity | 25 g |
Formula Weight | 238.45 |
Percent Purity | 97% |
Chemical Name or Material | 1-Chloro-3-iodobenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'