missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-hydroxypropane, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3757231
12.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-30-5
C3H7ClO
94.54
MFCD00002943
LAMUXTNQCICZQX-UHFFFAOYSA-N
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
12313
3-chloropropan-1-ol
C(CO)CCl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
627-30-5
C3H7ClO
94.54
MFCD00002943
LAMUXTNQCICZQX-UHFFFAOYSA-N
3-chloro-1-propanol, 3-chloropropanol, 1-propanol, 3-chloro, 1-chloro-3-hydroxypropane, trimethylene chlorohydrin, 3-chloro-l-propanol, 3-chlorpropan-1-ol, 3-choro-1-propanol, 1-chloro-3-propanol, 3-chloropropanol-1
12313
3-chloropropan-1-ol
C(CO)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -20°C
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 160°C to 162°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH2)3OH
Refractive Index 1.4440 to 1.446
Quantity 25 g
Beilstein 01, 356
Specific Gravity 1.13
Solubility Information Solubility in water: 300g/L water (20°C). Other solubilities: reacts with water forming hcl
Stabilizer 0.1% Na2CO3
Viscosity 6 mPa.s (20°C)
Formula Weight 94.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-3-hydroxypropane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-3-hydroxypropane, stabilized
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'