missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-nitrobenzene, 99+%, Acros Organics

£13.20 - £41.10

Chemical Identifiers

CAS 88-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD00007061
InChI Key BFCFYVKQTRLZHA-UHFFFAOYSA-N
Synonym 2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
PubChem CID 6945
ChEBI CHEBI:34270
IUPAC Name 1-chloro-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10030840 SDS View Documents Acros Organics
109621000
100mL £13.20
100mL View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
10713641 SDS View Documents Acros Organics
109620010
1L £41.10
1 L View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-06-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

88-73-3
157.56
BFCFYVKQTRLZHA-UHFFFAOYSA-N
6945
1-chloro-2-nitrobenzene
C6H4ClNO2
MFCD00007061
2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
CHEBI:34270
C1=CC=C(C(=C1)[N+](=O)[O-])Cl

Specifications

1-Chloro-2-nitrobenzene
124°C
Glass bottle
246°C
32°C to 34°C
99+%
99.0
99+%
ClC6H4NO2
05, 241
2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
BFCFYVKQTRLZHA-UHFFFAOYSA-N
1-chloro-2-nitrobenzene
6945
157.56
99+%
1.3480g/mL
Authentic
1.348
Yellow
100mL
88-73-3
100.0
C6H4ClNO2
MFCD00007061
15, 2153
Solubility in water: 0.43g/L water (20°C).,soluble in alcohol,benzene,ether,acetone,carbo,toluene,pyridine
C1=CC=C(C(=C1)[N+](=O)[O-])Cl
157.56
CHEBI:34270
Fused Crystalline Mass
Safety and Handling

Safety and Handling

Product Identifier
  • 1-Chloro-2-nitrobenzene

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.