missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpropane, 98%, Thermo Scientific Chemicals

Catalog Number p-7044037
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
513-36-0
C4H9Cl
92.566
MFCD00000954
QTBFPMKWQKYFLR-UHFFFAOYSA-N
10554
1-chloro-2-methylpropane
CC(C)CCl
This item is not returnable. View return policy
513-36-0
C4H9Cl
92.566
MFCD00000954
QTBFPMKWQKYFLR-UHFFFAOYSA-N
10554
1-chloro-2-methylpropane
CC(C)CCl

1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-methylpropane is used to prepare an organometllic compound, isobutyllithium by reacting with lithium using petroleum ether as a solvent.

Solubility
Miscible with water, alcohol and diethyl ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -131°C
Density 0.879
Boiling Point 68°C to 69°C
Flash Point −7°C (19°F)
Odor Sweet
Refractive Index 1.3975
Quantity 50 g
UN Number UN1127
Beilstein 635650
Merck Index 14,5138
Synonym isobutyl chloride, propane, 1-chloro-2-methyl, 1-chloro-2-methyl-propane, isobutylchlorid, iso-c4h9cl, sjphadikup@, acmc-1akdy, propane,1-chloro-2-methyl, ksc491k9j, 1-chloro-2-methylpropane
Solubility Information Miscible with water,alcohol and diethyl ether.
Formula Weight 92.57
Percent Purity 98%
Chemical Name or Material 1-Chloro-2-methylpropane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'