missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Chloro-2-(difluoromethoxy)-3-fluorobenzene, 97%

Brand:  Alfa Aesar™ H33151.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15495947

  • £33.50 / 1g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
196.55
MFCD22580772

Specifications

1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
1g
97%
MFCD22580772
Colorless
Liquid
196.55
Videos
SDS