missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Chloro-2-(difluoromethoxy)-3-fluorobenzene, 97%

Brand:  Alfa Aesar™ H33151.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15405957

  • £111.41 / 5g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
196.55
MFCD22580772

Specifications

1-Chloro-2-(difluoromethoxy)-3-fluorobenzene
5g
97%
MFCD22580772
Colorless
Liquid
196.55
Videos
SDS