1-Chloro-2,5-dibromobenzene, 99+%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 263410250

25GR 1-Chloro-2,5-dibromobenzene, 99+%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 3460-24-0 Weight: 0.07500kg


Packaging Glass bottle
Quantity 25g

Product Code. 10696524

Quantity Price
1 £ 31.7 / 25g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-Chloro-2, 5-dibromobenzene
Name Note 99+%
CAS 3460-24-0
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C6H3ClBr2
Formula Weight 270.34
MDL Number MFCD00018094
Synonym 1, 4-Dibromo-2-chlorobenzene

Specification

Assay ≥99.0%
Physical Form Fine Crystalline Powder
Formula Weight 270.34
Quantity 25g
Color Off-white
CAS Min % 99.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 3460-24-0
Infrared Spectrum Authentic
Assay Percent Range 99+%
MDL Number MFCD00018094
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 99+%
Molecular Formula C6H3ClBr2
CAS Max % 100.0
Chemical Name or Material 1-Chloro-2, 5-dibromobenzene
TSCA TSCA
Synonym 1, 4-Dibromo-2-chlorobenzene
Melting Point 39° to 42°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.