missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,5-dibromobenzene, 99+%, Thermo Scientific™

Catalog Number 10672403
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3460-24-0
C6H3ClBr2
270.34
MFCD00018094
LOWQAATYMJIWOG-UHFFFAOYSA-N
2-chloro-1,4-dibromobenzene, 2,5-dibromochlorobenzene, 1-chloro-2,5-dibromobenzene, benzene, 1,4-dibromo-2-chloro, pubchem3633, acmc-209i8v, chloro-3,6-dibrom obenzene, 1,4-bis bromanyl-2-chloranyl-benzene
2724603
1,4-dibromo-2-chlorobenzene
C1=CC(=C(C=C1Br)Cl)Br
This item is not returnable. View return policy
3460-24-0
C6H3ClBr2
270.34
MFCD00018094
LOWQAATYMJIWOG-UHFFFAOYSA-N
2-chloro-1,4-dibromobenzene, 2,5-dibromochlorobenzene, 1-chloro-2,5-dibromobenzene, benzene, 1,4-dibromo-2-chloro, pubchem3633, acmc-209i8v, chloro-3,6-dibrom obenzene, 1,4-bis bromanyl-2-chloranyl-benzene
2724603
1,4-dibromo-2-chlorobenzene
C1=CC(=C(C=C1Br)Cl)Br

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 39°C to 42°C
CAS Min % 99.0
Color White
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Fine Crystalline Powder
Assay Percent Range 99+%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 270.34
Percent Purity 99+%
Chemical Name or Material 1-Chloro-2, 5-dibromobenzene, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-2,5-dibromobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'