Learn More
Integra Biosciences™ 1-Channel Adapter
For use with VACUBOY Hand Operator.
Brand: Integra Biosciences™ 155522
Code : 04
Additional Details : Weight : 0.25000kg
Description
One-channel adaptor for use with VACUBOY hand operator.The VACUBOY hand operator can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
- With a precisely dosed volume
- Without loss of drops
- Without risk of contamination
- Without causing tiredness
- Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
- All parts in contact with fluids are autoclavable
Specifications
1-channel stainless steel adapter, 150 mm | |
VACUBOY Hand Operator |