missing translation for 'onlineSavingsMsg'
Learn More

1-Butylpyridinium bromide, 98%, Thermo Scientific™

Catalog Number p-80720
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
874-80-6
C9H14BrN
216.12
MFCD00671496
KVBQNFMTEUEOCD-UHFFFAOYSA-M
1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
2734238
1-butylpyridin-1-ium bromide
[Br-].CCCC[N+]1=CC=CC=C1
This item is not returnable. View return policy
874-80-6
C9H14BrN
216.12
MFCD00671496
KVBQNFMTEUEOCD-UHFFFAOYSA-M
1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
2734238
1-butylpyridin-1-ium bromide
[Br-].CCCC[N+]1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Ionic liquid
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 216.12
Percent Purity ≥97.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1-Butylpyridinium bromide
Show More Show Less
compliance-icons
Product Identifier
  • 1-Butylpyridinium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'