missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, ACS Reagent, ≥99.4%, Honeywell Riedel-de Haën™

Catalog Number 15635060
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
This item is not returnable. View return policy
71-36-3
C4H10O
74.123
MFCD00002964
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

ACS Reagent,≥99.4%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point -90°C
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Formula Weight 74.12g/mol
Percent Purity ≥99.4%
Density 0.81g/mL at 25°C
Boiling Point 116°C
Quantity 1 L
Explosion Limit 0.112
Flash Point 35°C (95°F)
Ignition Point 649°F
Infrared Spectrum Conforms to structure
Odor Characteristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.399
Chemical Name or Material 1-Butanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'