missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, 99+%, ACROS Organics™

Brand:  Acros Organics 220620010

Code : 29

Additional Details : CAS Number : 109-79-5 Weight : 0.89000kg Transport : UN number : 2347 Chem class : 1 Pack group : II

Quantity 1L
Packaging Glass bottle

 View more versions of this product

Product Code. 10616334

  • £49.60 / 1 L


EU Stock 35
Add to basket

Specifications

Chemical Identifiers

1-Butanethiol
109-79-5
100.0
99+%
CH3(CH2)3SH
MFCD00004905
99+%
99.0
≥99%
C4H10S
90.18
n-Butyl mercaptan

Specifications

1-Butanethiol
109-79-5
100.0
Glass bottle
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
≥99%
0.84
CH3(CH2)3SH
MFCD00004905
0.8400g/mL
15, 1580
− 116.0°C
98.0°C
0.5°C
99+%
99.0
Authentic
1.4420 to 1.4440
Liquid
99+%
C4H10S
90.18
01, 370
02,53; 04,64
Clear colorless
n-Butyl mercaptan
1L
Videos
SDS