missing translation for 'onlineSavingsMsg'
Learn More

1-Butanethiol, 98%, Thermo Scientific Chemicals

Catalog Number 10772885
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
109-79-5
C4H10S
90.18
MFCD00004905
WQAQPCDUOCURKW-UHFFFAOYSA-N
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
8012
butane-1-thiol
CCCCS
This item is not returnable. View return policy
109-79-5
C4H10S
90.18
MFCD00004905
WQAQPCDUOCURKW-UHFFFAOYSA-N
1-butanethiol, butanethiol, butyl mercaptan, n-butyl mercaptan, n-butanethiol, butylthiol, thiobutyl alcohol, n-butylmercaptan, 1-mercaptobutane, 1-butyl mercaptan
8012
butane-1-thiol
CCCCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -116.0°C
Color Colorless
Density 0.8400g/mL
Boiling Point 98.0°C
Flash Point 0.5°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)3SH
Refractive Index 1.4420 to 1.4440
Quantity 1 L
Beilstein 01, 370
Fieser 02,53; 04,64
Merck Index 15, 1580
Specific Gravity 0.84
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 90.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Butanethiol, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'