missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Bromopropane, 99%

Brand:  Alfa Aesar™ A10461.AU

UNSPSC : 12352101

Code : NEW

Additional Details : CAS Number : 106-94-5 Weight : 0.01000kg Transport : UN number : 2344 Chem class : 1 Pack group : II

Quantity 1000mL

 View more versions of this product

Product Code. 11438693

  • £38.16 / 1000mL


Estimated Shipment date
from Supplier 07-07-2017
Add to basket

Specifications

Description

Applications
1-Bromopropane is used as a solvent for adhesives, degreasing plastics and optics. It is used in the production of asphalt, synthetic fiber and aerosols. It is employed for cleaning the metal surfaces, electronic circuit boards, and multi-layered OLEDs (Organic Light Emitting Diodes). It serves to replace perchloroethylene as a dry cleaning solvent.

Solubility
Miscible with ethanol, water and diethyl ether.

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place.
Description & Specifications

Chemical Identifiers

1-Bromopropane
C3H7Br
123
UN2344
14,7845
106-94-5
CH3CH2CH2Br
MFCD00000254
505936
n-Propyl bromide

Specifications

1-Bromopropane
−110°C
1000mL
99%
CH3CH2CH2Br
MFCD00000254
505936
n-Propyl bromide
106-94-5
70° to 71°C
21°C (70°F)
C3H7Br
123
UN2344
14,7845
Documentation

Technical Documents