missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopinacolone, 97+%, Thermo Scientific Chemicals

Catalog Number p-7080915
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5469-26-1
C6H11BrO
179.057
MFCD00000206
SAIRZMWXVJEBMO-UHFFFAOYSA-N
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
21642
1-bromo-3,3-dimethylbutan-2-one
CC(C)(C)C(=O)CBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5469-26-1
C6H11BrO
179.057
MFCD00000206
SAIRZMWXVJEBMO-UHFFFAOYSA-N
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
21642
1-bromo-3,3-dimethylbutan-2-one
CC(C)(C)C(=O)CBr

1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.
TRUSTED_SUSTAINABILITY
Melting Point -10°C
Density 1.333
Boiling Point 189°C to 192°C
Flash Point 75°C (167°F)
Odor Pungent
Refractive Index 1.4655
Quantity 10 g
UN Number UN2810
Beilstein 506485
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 179.06
Percent Purity ≥97%
Chemical Name or Material 1-Bromopinacolone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'