missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctadecane, 96%, Thermo Scientific Chemicals

Catalog Number 10467750
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
112-89-0
C18H37Br
333.40
MFCD00000231
WSULSMOGMLRGKU-UHFFFAOYSA-N
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
8218
1-bromooctadecane
CCCCCCCCCCCCCCCCCCBr
This item is not returnable. View return policy
112-89-0
C18H37Br
333.40
MFCD00000231
WSULSMOGMLRGKU-UHFFFAOYSA-N
octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane
8218
1-bromooctadecane
CCCCCCCCCCCCCCCCCCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point >20.0°C
Color Yellow
Density 0.9700g/mL
Boiling Point 214.0°C to 216.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Linear Formula CH3(CH2)17Br
Refractive Index 1.4620 to 1.4640
Quantity 5 g
Specific Gravity 0.97
Formula Weight 333.38
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material 1-Bromooctadecane, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'