missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-2-(phenoxymethyl)benzene, 97%, Maybridge

Brand:  Maybridge CC58508EA

Code : 31

Additional Details : CAS Number : 861319-72-4 Weight : 0.06000kg Transport : UN number : 3261 Chem class : 1a Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10285904

  • £555.00 / 10g


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Specifications

Chemical Identifiers

1-(Bromomethyl)-2-(phenoxymethyl)benzene
96.5
97%
C14H13BrO
MFCD08690311
861319-72-4
100.0
97%
277.16

Specifications

1-(Bromomethyl)-2-(phenoxymethyl)benzene
96.5
Amber Glass Bottle
97%
C14H13BrO
MFCD08690311
861319-72-4
100.0
10g
97%
277.16
Videos
SDS